ADV509447712 AChemBlock

N,N-Dimethyl-2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]acetamide

  • MDL number MFCD18383279
  • Cas Number : 930596-20-6
  • Molecular Weight : 279.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/n-n-dimethyl-2-4-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrazol-1-yl-acetamide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13836
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart