ADV509447715 AChemBlock

3-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-4H,5H,6H-pyrrolo[1,2-b]pyrazole

  • MDL number MFCD22381490
  • Cas Number : 1314138-13-0
  • Molecular Weight : 234.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-tetramethyl-1-3-2-dioxaborolan-2-yl-4h-5h-6h-pyrrolo-1-2-b-pyrazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13840
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart