ADV509447716 AChemBlock

1-Methyl-1,3,8-triazaspiro[4.5]decan-4-one

  • MDL number MFCD20661682
  • Cas Number : 701897-99-6
  • Molecular Weight : 169.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-1-3-8-triazaspiro-4-5-decan-4-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13841
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart