ADV509447717 AChemBlock

Methyl 1H-pyrazolo[4,3-b]pyridine-7-carboxylate

  • MDL number MFCD22566131
  • Cas Number : 1363381-17-2
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-1h-pyrazolo-4-3-b-pyridine-7-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13842
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart