ADV509447718 AChemBlock

tert-Butyl N-[(1R,3R)-3-hydroxycyclopentyl]carbamate

  • MDL number MFCD19442807
  • Cas Number : 1290191-64-8
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-1r-3r-3-hydroxycyclopentyl-carbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13843
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart