ADV509447721 AChemBlock

(3R)-1-(Azetidin-3-yl)-3-fluoropyrrolidine dihydrochloride

  • MDL number MFCD23106353
  • Cas Number : 1403763-26-7
  • Molecular Weight : 217.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3r-1-azetidin-3-yl-3-fluoropyrrolidine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13846
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart