ADV509447725 AChemBlock

tert-Butyl 3-amino-3-(hydroxymethyl)azetidine-1-carboxylate

  • MDL number MFCD18375152
  • Cas Number : 1262411-27-7
  • Molecular Weight : 202.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-amino-3-hydroxymethyl-azetidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13850
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart