ADV509447728 AChemBlock

(4,4-Difluoro-1-methylcyclohexyl)methanamine

  • MDL number MFCD28502757
  • Cas Number : 1708157-79-2
  • Molecular Weight : 163.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-4-difluoro-1-methylcyclohexyl-methanamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13853
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart