ADV509447735 AChemBlock

(3S,5S)-5-Methylpyrrolidin-3-ol hydrochloride

  • MDL number MFCD19227362
  • Cas Number : 1107658-77-4
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3s-5s-5-methylpyrrolidin-3-ol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13861
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart