ADV509447736 AChemBlock

tert-Butyl 7-hydroxy-2-azaspiro[4.4]nonane-2-carboxylate

  • MDL number MFCD27986933
  • Cas Number : 1319716-41-0
  • Molecular Weight : 241.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-7-hydroxy-2-azaspiro-4-4-nonane-2-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13862
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart