ADV509447740 AChemBlock

Ethyl (1S,2S)-rel-2-(cyanomethyl)cyclopropane-1-carboxylate

  • MDL number MFCD18830853
  • Cas Number : 84673-47-2
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1s-2s-rel-2-cyanomethyl-cyclopropane-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13866
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart