ADV509447741 AChemBlock

(5S)-1,5-Dimethylpiperazin-2-one hydrochloride

  • MDL number MFCD23096888
  • Cas Number : 1887197-43-4
  • Molecular Weight : 164.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5s-1-5-dimethylpiperazin-2-one-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13867
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart