ADV509447743 AChemBlock

5-Bromo-6-chloro-1H-indazole-3-carboxylic acid

  • MDL number MFCD27987832
  • Cas Number : 1467062-18-5
  • Molecular Weight : 275.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-6-chloro-1h-indazole-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13869
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart