ADV509447746 AChemBlock

4-Amino-5-bromopyridine-2-carboxylic acid

  • MDL number MFCD21641994
  • Cas Number : 1363381-01-4
  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-amino-5-bromopyridine-2-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13873
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart