ADV509447749 AChemBlock

(S)-3,3-dimethylazetidine-2-carboxylic acid

  • MDL number MFCD30146439
  • Cas Number : 1860033-50-6
  • Molecular Weight : 129.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-azetidinecarboxylic-acid-3-3-dimethyl-2s.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13876
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart