ADV509447755 AChemBlock

Cis-2-{[(tert-butoxy)carbonyl]amino}cyclobutane-1-carboxylic acid

  • MDL number MFCD27665003
  • Cas Number : 951173-22-1
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/cis-2-tert-butoxy-carbonyl-amino-cyclobutane-1-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13882
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart