ADV509447761 AChemBlock

Methyl (1S,3R,4S)-3-{[(tert-butoxy)carbonyl]amino}-4-hydroxycyclopentane-1-carboxylate

  • MDL number MFCD02259718
  • Cas Number : 321744-14-3
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-1s-3r-4s-3-tert-butoxy-carbonyl-amino-4-hydroxycyclopentane-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13888
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart