ADV509447763 AChemBlock

3-Oxa-6-azabicyclo[3.1.1]heptane hydrochloride

  • MDL number MFCD30146441
  • Cas Number : 1860028-23-4
  • Molecular Weight : 135.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-oxa-6-azabicyclo-3-1-1-heptane-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13890
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart