ADV509447766 AChemBlock

Tert-butyl (3R,4R)-3-amino-4-fluoropyrrolidine-1-carboxylate

  • MDL number MFCD27918516
  • Cas Number : 1441392-27-3
  • Molecular Weight : 204.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3r-4r-3-amino-4-fluoropyrrolidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13893
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart