ADV509447767 AChemBlock

2-Methyl-2,5-diazabicyclo[2.2.2]octane dihydrochloride

  • MDL number MFCD26954798
  • Cas Number : 52321-18-3
  • Molecular Weight : 199.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-methyl-2-5-diazabicyclo-2-2-2-octane-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13894
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart