ADV509447770 AChemBlock

Tert-butyl 7-amino-2- azaspiro[4.4]nonane-2-carboxylate

  • MDL number MFCD24384500
  • Cas Number : 1341037-08-8
  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-7-amino-2-azaspiro-4-4-nonane-2-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13897
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart