ADV509447772 AChemBlock

2-{4-Aminobicyclo[2.2.2]octan-1-yl}acetic acid hydrochloride

  • MDL number MFCD30146442
  • Cas Number : 28345-44-0
  • Molecular Weight : 219.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-4-aminobicyclo-2-2-2-octan-1-yl-acetic-acid-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13899
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart