ADV509447775 AChemBlock

tert-Butyl 2-(8-bromo-7-methyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-1-yl)acetate

  • MDL number MFCD28501650
  • Cas Number : 1637533-84-6
  • Molecular Weight : 359.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-8-bromo-7-methyl-2-6-dioxo-2-3-6-7-tetrahydro-1h-purin-1-yl-acetate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13902
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart