ADV509447791 AChemBlock

tert-Butyl (2R,5R)-5-hydroxy-2-methylpiperidine-1-carboxylate

  • MDL number MFCD26793920
  • Cas Number : 1431473-05-0
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2r-5r-5-hydroxy-2-methylpiperidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13919
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart