ADV509447792 AChemBlock

1-(6-amino-3,5-difluoropyridin-2-yl)-7-(3-(3-((1-(6-amino-3,5-difluoropyridin-2-yl)-3-carboxy-8-chloro-6-fluoro-4-oxo-1,4-dihydroquinolin-7-yl)amino)-2-hydroxypropoxy)azetidin-1-yl)-8-chloro-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

  • MDL number MFCD28502726
  • Cas Number : 1093185-35-3
  • Molecular Weight : 881.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-quinolinecarboxylic-acid-1-6-amino-3-5-difluoro-2-pyridinyl-7-3-3-1-6-amino-3-5-difluoro-2-pyridinyl-3-carboxy-8-chloro-6-fluoro-1-4-dihydro-4-oxo-7-quinolinyl-amino-2-hydroxypropoxy-1-azetidinyl-8-chloro-6-fluoro-1-4-dihydro-

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13920
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart