ADV509447844 AChemBlock

2-[(3S)-1-[(tert-Butoxy)carbonyl]pyrrolidin-3-yl]acetic acid

  • MDL number MFCD05861546
  • Cas Number : 204688-61-9
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-3s-1-tert-butoxy-carbonyl-pyrrolidin-3-yl-acetic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13977
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart