ADV509447862 AChemBlock

[4-(Hydroxymethyl)pyridin-3-yl]methanol hydrochloride

  • MDL number MFCD29918533
  • Cas Number : 53654-42-5
  • Molecular Weight : 175.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-hydroxymethyl-pyridin-3-yl-methanol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13996
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart