ADV509447875 AChemBlock

Ethyl 2-bromo-1,3-oxazole-4-carboxylate

  • MDL number MFCD09801023
  • Cas Number : 460081-20-3
  • Molecular Weight : 220.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-bromo-1-3-oxazole-4-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14010
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart