ADV509447877 AChemBlock

Cyclopentylmethanamine

  • MDL number MFCD04114081
  • Cas Number : 6053-81-2
  • Molecular Weight : 99.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/cyclopentylmethanamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14012
pictograms GHS02; GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart