ADV509447889 AChemBlock

Di-tert-Butyl chloromethyl phosphate

  • MDL number MFCD12165895
  • Cas Number : 229625-50-7
  • Molecular Weight : 258.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/di-tert-butyl-chloromethyl-phosphate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14022
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart