ADV509447900 AChemBlock

4-Bromo-3-iodo-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD09880123
  • Cas Number : 1000340-34-0
  • Molecular Weight : 322.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-3-iodo-1h-pyrrolo-2-3-b-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14033
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart