ADV509447905 AChemBlock

4-(Cyclohex-1-en-1-yl)-3-(trifluoromethyl)benzoic acid

  • MDL number MFCD30146450
  • Cas Number : 1034188-31-2
  • Molecular Weight : 270.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-cyclohex-1-en-1-yl-3-trifluoromethyl-benzoic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14038
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart