ADV509447906 AChemBlock

5-Bromo-4-chloropyrimidin-2-amine

  • MDL number MFCD11046979
  • Cas Number : 1044767-99-8
  • Molecular Weight : 208.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-4-chloropyrimidin-2-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14039
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart