ADV509447912 AChemBlock

(1R,2S)-2-Fluorocyclopropan-1-amine; 4-methylbenzene-1-sulfonic acid

  • MDL number MFCD01861147
  • Cas Number : 143062-84-4
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1r-2s-2-fluorocyclopropan-1-amine-4-methylbenzene-1-sulfonic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14045
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart