ADV509447913 AChemBlock

2-Chloro-1H,4H-pyrrolo[2,1-f][1,2,4]triazin-4-one

  • MDL number MFCD29060177
  • Cas Number : 1613751-69-1
  • Molecular Weight : 169.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-1h-4h-pyrrolo-2-1-f-1-2-4-triazin-4-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14046
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart