ADV509447941 AChemBlock

Benzyl 4-hydroxy-4-(trifluoromethyl)piperidine-1-carboxylate

  • MDL number MFCD19686795
  • Cas Number : 946504-83-2
  • Molecular Weight : 303.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/benzyl-4-hydroxy-4-trifluoromethyl-piperidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14075
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart