ADV509447944 AChemBlock

Bis(2-azabicyclo[2.2.1]hept-5-ene); oxalic acid

  • MDL number MFCD29918564
  • Cas Number : 1860028-31-4
  • Molecular Weight : 280.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/bis-2-azabicyclo-2-2-1-hept-5-ene-oxalic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14078
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart