ADV509447945 AChemBlock

6-Chloropyridine-3,4-diamine

  • MDL number MFCD04037269
  • Cas Number : 89182-17-2
  • Molecular Weight : 143.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-chloropyridine-3-4-diamine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14079
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart