ADV509447949 AChemBlock

tert-butyl 3-hydroxy-3,6-dihydropyridine-1(2H)-carboxylate

  • MDL number MFCD04972246
  • Cas Number : 224779-27-5
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-2h-pyridinecarboxylic-acid-3-6-dihydro-3-hydroxy-1-1-dimethylethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14083
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart