ADV509447959 AChemBlock

2-(Trifluoromethyl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD18632534
  • Cas Number : 1286734-84-6
  • Molecular Weight : 197.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-trifluoromethyl-1-3-thiazole-5-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14093
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart