ADV509447963 AChemBlock

4-Bromo-2-iodo-5-methylaniline

  • MDL number MFCD27995559
  • Cas Number : 1609264-62-1
  • Molecular Weight : 311.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-2-iodo-5-methylaniline.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14097
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart