ADV509447977 AChemBlock

5-Bromopyrazolo[1,5-a]pyridine-3-carboxylic acid

  • MDL number MFCD18837595
  • Cas Number : 1101121-05-4
  • Molecular Weight : 241.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromopyrazolo-1-5-a-pyridine-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14111
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart