ADV509447981 AChemBlock

2-Chloro-3H,4H,7H-pyrrolo[2,3-d]pyrimidin-4-one

  • MDL number MFCD23702052
  • Cas Number : 1245811-22-6
  • Molecular Weight : 169.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-3h-4h-7h-pyrrolo-2-3-d-pyrimidin-4-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14115
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart