ADV509447988 AChemBlock

2,2-Dimethylmorpholine hydrochloride

  • MDL number MFCD16037342
  • Cas Number : 167946-94-3
  • Molecular Weight : 151.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-2-dimethylmorpholine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14122
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart