ADV509448010 AChemBlock

5-Bromo-2-chloro-N-(prop-2-en-1-yl)pyrimidin-4-amine

  • MDL number MFCD29918560
  • Cas Number : 959799-08-7
  • Molecular Weight : 248.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-chloro-n-prop-2-en-1-yl-pyrimidin-4-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14144
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart