ADV509448016 AChemBlock

6-Chloro-4-iodopyridine-3-carbonitrile

  • MDL number MFCD16658125
  • Cas Number : 1061357-83-2
  • Molecular Weight : 264.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-4-iodopyridine-3-carbonitrile.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14150
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart