ADV509448027 AChemBlock

1-[(5-Chloro-1H-indol-3-yl)methyl]piperidine-4-carboxylic acid

  • MDL number MFCD14281859
  • Cas Number : 1170595-16-0
  • Molecular Weight : 292.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-5-chloro-1h-indol-3-yl-methyl-piperidine-4-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14161
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart