ADV509448035 AChemBlock

Ethyl 2-(5-chloro-1H-indol-3-yl)-2-oxoacetate

  • MDL number MFCD12546261
  • Cas Number : 163160-58-5
  • Molecular Weight : 251.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-5-chloro-1h-indol-3-yl-2-oxoacetate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14169
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart