ADV509448061 AChemBlock

tert-Butyl (2R,4R)-4-hydroxy-2-methylpyrrolidine-1-carboxylate

  • MDL number MFCD11109735
  • Cas Number : 114676-93-6
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2r-4r-4-hydroxy-2-methylpyrrolidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14196
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart