ADV509448064 AChemBlock

(1,2-Oxazol-3-yl)methanamine hydrochloride

  • MDL number MFCD06738790
  • Cas Number : 1187933-48-7
  • Molecular Weight : 134.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-2-oxazol-3-yl-methanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14199
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart